Would you like to log in again?
Pierce™ Protein L Plus Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
20520 20520
PI20520EA 622.52 USD InStock
PI20520 CAPI20520
Pierce™ Protein L Plus Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Protein L Plus agarose is a high-capacity Protein L beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein L is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non-specific binding and enabling multiple uses without decline in yield
  • High capacity beads with a dense load of immobilized Protein L
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty