Would you like to log in again?
Restore™ Western Blot Stripping Buffer, Thermo Scientific
Supplier: Thermo Scientific
21062 21059 21059 21063 21063 21062
PI21059EA 210.02 USD InStock
CAPI21063 PI21059 CAPI21062 PI21063 PI21062 CAPI21059
Restore™ Western Blot Stripping Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Stripping Buffers
Thermo Scientific Restore™ Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
  • Saves time—no need to re-run gels and blots
  • Saves costly sample—re-probe the membrane using the same target sample
  • Effective—formulation is more efficient at stripping antibodies than homemade buffers
  • Gentle—does not damage the target antigen during stripping allowing efficient reprobing
  • Odor-free—no mercaptans means no acrid odors
  • Economical—less expensive than other commercial stripping buffers
Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore™ Western blot stripping buffer offers an effective alternative.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at room temperature or 4˚C. Product shipped at ambient temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty