Would you like to log in again?
Pierce™ Streptavidin, Maleimide-Activated, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
21102 21102
PI21102EA 340.29 USD InStock
PI21102 CAPI21102
Pierce™ Streptavidin, Maleimide-Activated, Thermo Scientific
Protein Purification Protein Purification Reagents
Thermo Scientific Pierce Maleimide-Activated Streptavidin conjugate include streptavidin in a purified form, activated for crosslinking to sulfhydryl groups containing molecules.
  • Provided as a lyophilized powder containing sodium citrate
  • Facilitates conjugation of Streptavidin to proteins
Maleimide-activated Streptavidin is designed to facilitate the conjugation of the Streptavidin to proteins, peptides and ligands that contain sulfhydryl (-SH) groups. The reactive maleimide group that has been added to the Streptavidin allows conjugation reactions to be carried out in the pH range 6.5-7.5, creating a stable thio-ether link.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty