Would you like to log in again?
Recombinant Streptavidin, HRP (Horseradish Peroxidase), Thermo Scientific
Supplier: Thermo Scientific
21124
PI21124EA 573.78 USD InStock
PI21124
Recombinant Streptavidin, HRP (Horseradish Peroxidase), Thermo Scientific
Proteins and Peptides
Use to develop in-house non-radioactive assay systems.
Avidin can be used in ELISAs, immunohistochemical staining, Western blotting, hybridization, affinity chromatography, and fluorescent-activated cell sorting. The streptavidin conjugates are recommended for use where high binding affinity and specificity is required. NeutrAvidin® biotin-binding protein is a more economical alternative to both avidin and streptavidin. It features low nonspecific binding properties with no glycosylation. It also offers a neutral pI and lysine residues that remain available for derivatization or conjugation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty