Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Recombinant Protein A/G

Supplier: Pierce Chemical
Pierce
77678 21186 21186 77677 77678 77677
PI77677EA 729.23 USD InStock
CAPI77677 CAPI21186 PI77677 CAPI77678 PI77678 PI21186
Pierce™ Recombinant Protein A/G
Proteins and Peptides
Purified (unconjugated) Thermo Scientific Pierce Recombinant Protein A/G is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of rabbit and human antibodies, especially IgG isotypes, in immunoassays and antibody purification protocols.

  • Contains four Fc-binding domains from Protein A and two from Protein G
  • Provides binding for all antibody species and subclasses recognised by either Protein A or Protein G

The extended Fc-binding properties of Protein A/G make it a tool in the investigation and purification of immunoglobulins. Protein A/G binds to all human IgG subclasses, IgA, IgE, IgM and to a lesser extent IgD; however, it does not bind mouse IgA, IgM or murine serum albumin. Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins. Individual subclasses of mouse monoclonal are most likely to have stronger affinity to this chimeric protein than to either Protein A or Protein G.

Properties of Recombinant Protein A/G:
Source: E. coli
Molecular weight: 50,460 (Apparent MW by SDS-PAGE: 40,000 to 45,000)
Form: salt-free powder
A280 of 0.1% solution: 0.505
Isoelectric point (pI): 4.65

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty