Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Recombinant Protein G

Supplier: Pierce Chemical
Pierce Pierce™
77676 21193 77675 77676 77675 21193
PI21193EA 266.95 USD InStock
CAPI21193 CAPI77676 CAPI77675 PI21193 PI77676 PI77675
Pierce™ Recombinant Protein G
Proteins and Peptides
Purified (unconjugated) Thermo Scientific Pierce™ Recombinant Protein G is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of rabbit and human antibodies, especially IgG isotypes, in immunoassays and antibody purification protocols.

  • Contains two Fc-binding domains per protein
  • Does not bind human IgM, IgD or IgA
  • More efficient than protein A for mouse (IgG1), human (IgG3) and rat, goat and cow antibodies
  • Less efficient than protein A for guinea pig, pig, dog and cat

Protein G is a bacterial cell wall protein isolated from group G Streptococci. DNA sequencing of native protein G identifies two IgG-binding domains and sites for albumin and cell surface binding. The albumin and cell surface binding domains have been eliminated from recombinant protein G to reduce non specific binding and, therefore, can be used to separate IgG from crude samples. Optimal binding occurs at pH 5, although binding is also effective at pH 7.0 to 7.2.

Properties of Recombinant Protein G:
Source: E. coli
Molecular Weight:

21,600 (Apparent MW by SDS-PAGE: 32,000)
Form: salt-free powder
A280 of 0.1% solution: 1.0
Isoelectric point (pI): 4.5

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty