Would you like to log in again?
Pierce™ Protein A Plus Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
22810 22811 22811 22810 22812 22812
PI22810EA 175.02 USD InStock
CAPI22811 CAPI22810 PI22810 PI22811 CAPI22812 PI22812
Pierce™ Protein A Plus Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Protein A Plus agarose is a high capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • High capacity beads with a dense load of immobilized Protein A
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty