Would you like to log in again?
Pierce™ Dye Removal Columns, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
22858 22858
PI22858EA 234.95 USD InStock
CAPI22858 PI22858
Pierce™ Dye Removal Columns, Thermo Scientific
Protein Linking and Labeling Systems
Pierce™ Dye Removal Columns enable fast and efficient removal of non-reacted fluorescent dyes from protein labeling reactions.
  • Removes free, unconjugated dye from labeled protein solutions in less than a minute
  • Purification resin provides outstanding conjugate recovery
  • Separate resin and microcentrifuge columns allow optimization for different dyes and concentrations of sample
Removing excess dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. The dye removal resin in this kit is highly specialized to produce exceptional protein recoveries while effectively removing non-conjugated dye. Using the appropriate amount of resin and buffer conditions, almost any fluorescent dye can be removed with this kit.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty