Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Pierce™

Supplier: 0000013509
Warning
Synonyms: EDAC hydrochlorideEDAC HClEDC HClEDC1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochlorideWSC HCl1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochlorideN-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochlorideEDC hydrochlorideN'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride

22980 77149 22981 22981 77149 22980
PI22980EA 175.32 USD InStock
PI22980 PI22981 CAPI77149 PI77149 CAPI22981 CAPI22980
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Pierce™
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Thermo Scientific Pierce EDC is a carboxyl- and amine-reactive zero-length crosslinker. EDC reacts with a carboxyl group first and forms an amine-reactive O-acylisourea intermediate that quickly reacts with an amino group to form an amide bond with release of an isourea by-product. The intermediate is unstable in aqueous solutions and so two-step conjugation procedures rely on N-hydroxysuccinimide (NHS) for stabilization. Failure to react with an amine will result in hydrolysis of the intermediate, regeneration of the carboxyl, and release of an N-substituted urea. A side reaction is the formation of an N-acylurea, which is usually restricted to carboxyls located in hydrophobic regions of proteins.

  • Reactive group: carbodiimide
  • Reaction target: activates carboxyl groups to conjugate to amino groups (primary amines)
  • Several conjugation strategies—react EDC alone with target groups or include NHS or Sulfo-NHS to increase reaction efficiency or to stabilize active intermediate for later reaction to amines
  • Neutral linkage—forms neutral amide bonds between carboxyls and amines
  • Water-soluble reagent—add directly to reactions in aqueous, physiological buffers
  • Soluble reaction byproducts—easily removed by washing with water or dilute acid
  • High-purity, crystalline reagent—use to create high-quality, activated derivatives

1-Ethyl-3-[3-dimethylaminopropyl]carbodiimide hydrochloride (EDC or EDAC) is a zero-length crosslinking agent used to couple carboxyl groups to primary amines. This crosslinker has been used in diverse applications such as forming amide bonds in peptide synthesis, attaching haptens to carrier proteins to form immunogens, labeling nucleic acids through 5' phosphate groups and creating amine-reactive NHS-esters of biomolecules. EDC reacts with a carboxyl to form an amine-reactive O-acylisourea intermediate. If this intermediate does not encounter an amine, it will hydrolyze and regenerate the carboxyl group. In the presence of N-hydroxysulfosuccinimide (Sulfo-NHS), EDC can be used to convert carboxyl groups to amine-reactive Sulfo-NHS esters. This is accomplished by mixing the EDC with a carboxyl containing molecule and adding Sulfo-NHS.


Formula: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Melting Pt: ∼ 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012503
CAS Number: 25952-53-8
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/32/20961432.jpg
Order Now
Specification Test Results
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short
Form Powder
Quantity 10 mg/5 g/25 g
Type Crosslinker
Solubility Water

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty