Would you like to log in again?
Pierce™ Coomassie (Bradford) Protein Assay Kits, Thermo Scientific
Supplier: Thermo Scientific
Pierce
23200 23200
PI23200EA 175.32 USD InStock
PI23200 CAPI23200
Pierce™ Coomassie (Bradford) Protein Assay Kits, Thermo Scientific
Assays Protein Assays
The Pierce™ Coomassie (Bradford) Protein Assay Kit is a ready-to-use, stable formulation of the traditional Bradford assay reagent to measure (A595 nm) total protein concentration compared to a protein standard.
  • Stable, ready-to-use kit of the classical Bradford assay reagent
  • Colorimetric measure with a standard spectrophotometer or plate reader (595 nm)
  • Easy to use, only single reagent; no working reagent preparation required
  • Almost immediate color development; add, mix, and read results
  • Detects protein concentration in the range 1 to 1,500 µg/mL
  • Microplate and cuvette protocols provided with the instructions and adaptable to several target working ranges
When Coomassie dye binds protein in an acidic medium, an immediate shift in absorption maximum occurs from 465nm to 595nm with a concomitant colour change from brown to blue.
Delivery information: Kit contains Pierce™ Coomassie Protein Assay Reagent (950 mL); Albumin Standard Ampoules (2 mg/mL,10x1 mL).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty