Would you like to log in again?
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent, Thermo Scientific
Supplier: Thermo Scientific
Pierce
23238 23238
PI23238EA 228.63 USD InStock
PI23238 CAPI23238
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent, Thermo Scientific
Assays Protein Assays
The Bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration
  • Sensitive, fast and consistent protein assay kit
  • Determine protein concentrations from 1 to 1500 μg/mL
  • Convenient microplate or cuvette format
  • Compatible with reducing sugars, reducing substances and thiols
This modification of the well-known Bradford method greatly reduces the tendency of Coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, Coomassie Plus Reagent results in significantly less protein-to-protein variation than is observed with other Bradford-type Coomassie formulations.
The Pierce™ Coomassie Plus Protein Assay is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent to quickly measure (A595 nm) total protein concentration compared to a protein standard.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty