Would you like to log in again?
Pierce™ Compat-Able™ Coomassie Plus Protein Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
23239 23239
PI23239EA 430.02 USD InStock
PI23239 CAPI23239
Pierce™ Compat-Able™ Coomassie Plus Protein Assay Kit, Thermo Scientific
Assays Protein Assays
The Compat-Able™ Coomassie Plus Protein Assay Kit eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
  • Eliminate incompatibilities, removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
  • Simple and fast, prepare samples in less than 10 minutes with four simple steps
  • Reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
  • Microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
  • Tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalisation procedures
The Compat-Able™ reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods.
Delivery information: Kit contains Compat-Able™ Reagent 1 (250 mL); Compat-Able™ Reagent 2 (250 mL); Pierce™ Coomassie Plus Assay Reagent (950 mL); Albumin Standard Ampoules (2 mg/mL, 10x1 mL).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty