Would you like to log in again?
Pierce™ Modified Lowry Protein Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
23240 23240
PI23240EA 264.17 USD InStock
CAPI23240 PI23240
Pierce™ Modified Lowry Protein Assay Kit, Thermo Scientific
Assays Protein Assays
The Modified Lowry Protein Assay Kit is the preferred colorimetric assay for quantifying total protein concentration.
  • Ready-to-use reagent for the loyal Lowry method user
  • Preformulated cupric sulfate-tartrate reagent stable for one year at room temperature
  • Linear results from 1 to 1,500 µg/mL for BSA
  • Adaptable to microplates
  • Less protein:protein variation than dye-binding methods
The sensitivity of the Modified Lowry Protein Assay is greatly enhanced over that of the biuret reagent. The working range of the method extends from 5 to 2,000 mg/mL. Using cold Modified Lowry Protein Assay Reagent will result in low absorbance values.
Delivery information: Kit includes Modified Lowry Protein Assay Reagent (480 mL), 2 N Folin-Ciocalteu Phenol Reagent (50 mL), Albumin Standard Ampoules, 2 mg/mL, (10×1 mL).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty