Would you like to log in again?
Pierce™ Detergent Compatible Bradford Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
23246
PI23246EA 261.31 USD InStock
PI23246
Pierce™ Detergent Compatible Bradford Assay Kit, Thermo Scientific
Assays Protein Assays
The Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Kit is a quick and ready-to-use modification of the well-known Bradford coomassie dye-binding, colorimetric method for total protein quantitation
Proprietary additives to the Bradford Reagent make it compatible with up to 1% or higher of detergents and lysis reagents that are commonly used in life science research, including Triton® X-100 and NP-40.Convenient— detergent-free standard curveFlexible— compatible with samples both with and without detergentMinimal sample—requires only 10 µL for microplate procedureColorimetric—measure with a standard spectrophotometer or plate reader (595 nm)Easy to use— single reagent, no working reagent preparation requiredFast—10 minute incubation at room temperatureBroad range—detects protein concentration in the ranges of 2 to 1500 µg/mLSimilar to the Bradford method, coomassie dye binds protein in an acidic medium causing an immediate shift in absorption maximum from 465 nm to 595 nm with a concomitant color change from green to blue. In addition, the assay is complete in just 10 minutes.The protein assay can be performed in either test tube or microplate format. The standard working range is 100-1500 µg/mL with up to 1% detergent (or higher in some cases). Protein concentrations are estimated by reference to absorbances obtained for a series of standard protein dilutions, typically bovine serum albumin (BSA), which are assayed alongside the unknown samples. Because the color response with coomassie dye is non-linear with increasing protein concentration, a standard curve must be completed with each assay. Standards can be used directly without preparing them in the same detergent found in the test samples.
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Detergent Compatible Bradford Assay Reagent, 450 mL, store at 4˚C
Albumin Standard Ampules, 10 x 1 mL, store at room temperature
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty