Would you like to log in again?
Pierce PeroXOquant™ Quantitative Peroxide Assay Kits (Lipid), Thermo Scientific
Supplier: Thermo Scientific
Pierce PeroXOquant™
23285 23285
PI23285EA 273.97 USD InStock
CAPI23285 PI23285
Pierce PeroXOquant™ Quantitative Peroxide Assay Kits (Lipid), Thermo Scientific
Assays Protein Assays
Pierce™ Quantitative Peroxide Assay Kit (Lipid) detects and measures hydrogen peroxide levels in biological samples using an Iron and Xylenol orange* (XO) reagent.
  • Ready to use – removes the variability, difficulty and expense of formulating in-house
  • Ideal for sensitive measurement of peroxide accumulation in non-lipid samples
This Lipid-compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange
Peroxides in the sample oxidize Fe2+ to Fe3+. The Fe3+ ion will form a colored complex with xylenol orange that can be read at 560nm.
*Xylenol orange is (o-cresolsulfonephthalein-3´-3´-bis-[methyliminodiacetic acid sodium salt]).
It can be used with lipid-containing samples without extracting the lipids. The assay is less sensitive, since it does not contain sorbitol. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (ie. control) to mitigate these effects.
Delivery information: Complete kit includes Reagent A (1 mL) and Reagent C (4x25 mL).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty