Would you like to log in again?
Pierce Easy-Titer® IgM Assay Kits, Thermo Scientific
Supplier: Thermo Scientific
Pierce Easy-Titer®
23315 23315
PI23315EA 923.08 USD InStock
PI23315 CAPI23315
Pierce Easy-Titer® IgM Assay Kits, Thermo Scientific
Assays Immunological Assays
Easy-Titer® IgM Assay Kit is a simple, mix-and-read assay that allows quick and accurate determination of concentrations from 15 to 500 ng/mL of IgM
  • Faster and easier than ELISA
  • Sensitive, requires small sample volume
  • No special equipment needed
  • Highly reproducible results
The assay procedure uses monodispersed polystyrene beads that are coated with anti-human IgM antibodies and absorb light at 340 and 405 nm. When the beads are mixed with a sample containing human IgM, they aggregate, causing decreased absorption of light and, therefore, low IgM concentrations yield high absorbance values and high IgM concentrations yield low absorbance values. The decrease in absorption is proportional to IgM concentration and a standard curve can be generated to accurately quantify IgM levels in serum or citrated plasma samples.
Delivery information: Kit include sufficient components for 96 tests. An IgM standard must be purchased separately. Each kit contains goat Anti-Human IgM Coated polystyrene beads (2 mL), Dilution buffer (30 mL), and blocking reagent (15 mL).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty