Would you like to log in again?
Pierce ProFound™ Co-Immunoprecipitation Kits, Thermo Scientific
Supplier: Thermo Scientific
Pierce ProFound™
23625 23620 23620 23625
PI23625EA 585.85 USD InStock
CAPI23625 PI23625 PI23620 CAPI23620
Pierce ProFound™ Co-Immunoprecipitation Kits, Thermo Scientific
Assays Protein Assays
An ideal system for designing, constructing, and performing a controlled co-immunoprecipitation (co-IP) experiment.
  • Immobilized anti-c-Myc monoclonal antibody binds the c-Myc epitope tag with high specificity, providing high yield immunoprecipitation products and clean Western blot detection
  • Excellent results with as little as 1 µg of anti-c-Myc antibody in IP mode with the positive control lysate
  • Kit includes a positive control lysate that contains over-expressed GST-c-Myc to validate the reagents and specific protocol, and the instructions provide tips for planning other controls
  • Affinity system is compatible with IP or Co-IP from various cell lysates and physiological (non-denaturing) buffer systems
  • Complete kit includes all necessary reagents, convenient spin columns, and easy-to-follow instructions
Kits use AminoLink® coupling gel to immobilize a purified antibody and retain antibody function
The HA and c-Myc tagged protein kits feature a high-affinity antibody for high yield and minimal background. These kits are also scalable for different antibody-gel and lysate requirements. Mammalian kits include M-PER™ mammalian protein extraction reagent for easy extract preparation that is compatible with the co-IP protocols.
In addition, the antibody-coupled core support retains the antibody during elution of the co-IP complex, eliminating interference from antibody fragments during detection of the co-IP products. Because the antibody is attached covalently, the antibody-gel matrix can be reused up to 10 times, conserving antibody. Kits will work with any antibody regardless of species or Ig subclass. The protocols are designed for spin columns for efficient washing and elution of protein samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty