Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ HisPur™ Ni-NTA Superflow Agarose, Thermo Scientific

Supplier: Thermo Fisher Scientific
HisPur™
25215 25217 25214 25215 25216 25217 25214 25216
PI25217EA 10578.62 USD InStock
PI25217 PI25214 PI25216 PI25215 CAPI25214 CAPI25217 CAPI25216 CAPI25215
Pierce™ HisPur™ Ni-NTA Superflow Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce HisPur™ Ni-NTA Superflow Agarose is a nitrilotriacetic acid modified Superflow 6 support charged with divalent nickel (Ni+2) used for FPLC purification of poly-histidine-tagged proteins.

  • Binds greater than 20 mg of His-tagged GFP per ml of resin at a linear flow rate of 300 cm/hour
  • Provides greater than 80% purity of eluted fractions when used to purify from lysates
  • Can be used to purify proteins under native and denaturing conditions
  • Highly crosslinked beads tolerate linear flow rates up to 1200 cm/hour
  • Maintains function in a wide variety of chemicals and pH conditions
  • Resin is stable through multiple cycles of cleaning and reuse

HisPur™ Ni-NTA Superflow Agarose is a durable resin that does not compress at flow rates used for medium- or large-scale FPLC purifications.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty