Would you like to log in again?
Pierce™ HisPur™ Cobalt Superflow Agarose, Thermo Scientific
Supplier: Thermo Fisher Scientific
HisPur™
25229 25231 25230 25228 25230 25229 25228 25231
PI25229EA 689.45 USD InStock
PI25229 PI25228 CAPI25231 CAPI25230 PI25231 CAPI25229 CAPI25228 PI25230
Pierce™ HisPur™ Cobalt Superflow Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce HisPur™ Cobalt Superflow Agarose is a tetradentate-chelating Superflow 6 support charged with divalent cobalt, designed for high-purity, large-scale purification of polyhistidine-tagged proteins.
  • Binds 20 mg of 6xHis-GFP per ml of resin at a linear flow rate of 150 cm/hour; can bind >30 mg per mL of resin depending on flow rate
  • Highly crosslinked beads tolerate linear flow rates up to 1200 cm/hour with minimal leaching of cobalt
  • Specificity of cobalt binding to histidine-tag generally results in elution fractions with less than 10% contamination of non specific proteins
  • Maintains function after exposure to a wide variety of chemicals and pH values
  • Resin is stable through multiple cycles of cleaning and reuse
HisPur™ Cobalt Superflow Agarose is a durable resin that does not compress at flow rates used for medium- or large-scale FPLC purifications.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty