Would you like to log in again?
Pierce™ Glutathione Superflow Agarose Affinity Chromatography Media, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
25238 25238 25236 25237 25237 25236
PI25236EA 342.35 USD InStock
PI25236 PI25238 PI25237 CAPI25236 CAPI25238 CAPI25237
Pierce™ Glutathione Superflow Agarose Affinity Chromatography Media, Thermo Scientific
Protein Purification Protein Purification Resins
Thermo Scientific Pierce Glutathione Superflow Agarose consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose that is designed for high-purity, large-scale purification of GST-tagged proteins.
  • High dynamic binding capacity—binds 10 mg of pure GST per mL of resin at a linear flow rate of 150 cm/hour; can bind greater than 20 mg/mL at 30 cm/hour flow rate
  • High purity—provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
  • Robust—highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200 cm/hour
  • Compatible—maintains function after exposure to a wide variety of chemicals and pH values
  • Cost effective—resin is stable through multiple cycles of cleaning and reuse
Glutathione Superflow Agarose is a highly crosslinked, durable resin that does not compress at flow rates required for medium- to large-scale FPLC purifications. The resin holds up well to a variety of chemical and pH values, and it is compatible with common clean-in-place procedures. The specific binding of glutathione to the GST-tag results in high purity and high yield in a single step purification process.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty