Would you like to log in again?
Pierce™ Pegylation Reagents, Methyl-PEG-Amine Compounds, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
26112 26113 26110 26117 26116 26111 26115 26112 26113 26111 26110 26115 26114 26116 26114 26117
PI26117EA 2114.46 USD InStock
CAPI26112 PI26117 CAPI26111 CAPI26110 PI26115 PI26116 PI26113 CAPI26116 CAPI26115 PI26114 PI26111 CAPI26114 CAPI26113 PI26112 PI26110 CAPI26117
Pierce™ Pegylation Reagents, Methyl-PEG-Amine Compounds, Thermo Scientific
Protein Linking and Labeling Systems
The methyl-PEGn-amine (MA[PEG]n) PEGylation reagents are methyl ether-terminated PEG amines that are used for modifying proteins or surfaces such as beads, nanoparticles and self-assembled monolayers
  • Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application
  • PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Allows site-specific labeling of primary amines or carboxyl groups on proteins or surfaces
  • Easy-to-follow instructions increase the likelihood of a successful outcome
MA(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers and terminal methyl (-CH3) and amino (-NH2) groups. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-Amine, where the subscript "n" denotes 4, 8, 12, or 24 ethylene glycol units. The terminal primary amine of each compound provides a specific target for crosslinking and other conjugation methods, making these compounds useful as PEGylation reagents.
These PEGylation reagents are homogenous compounds of defined molecular weight and spacer length, providing precision in optimizing modification applications.
Ordering information: Upon receipt store product at -20?C protected from moisture. Product is shipped at ambient temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty