Would you like to log in again?
Pierce® Pegylation Reagents, Methyl-PEG-Thiol Compound, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce® Pierce™
26132 26132
PI26132EA 422.58 USD InStock
CAPI26132 PI26132
Pierce® Pegylation Reagents, Methyl-PEG-Thiol Compound, Thermo Scientific
Protein Purification Protein Purification Reagents
Methyl-PEG4-thiol is a methyl- and sulfhydryl-terminated compound that contains a 4-unit polyethylene glycol (PEG) spacer and is used to modify surfaces such as quantum dots, monolayers and magnetic particles.
  • PEG groups are flexible, non-immunogenic and hydrophilic
  • Terminal monodentate thiol reacts spontaneously with gold surfaces through dative binding
  • Terminal methyl group coats quantum dot and other surfaces to reduce nonspecific binding in assays
The use of MT(PEG)4 with CT(PEG)12 in surface modification can form a hydrophilic 'lawn' of methyl ether-terminated PEGs with periodic exposed carboxylic acid-containing PEGs. The exposed carboxylic acid groups can be coupled to affinity ligands using the carbodiimide coupling reaction with EDC and sulfo-NHS.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty