Would you like to log in again?
Pierce™ Agarose ChIP Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
26156 26156
PI26156EA 925.29 USD InStock
CAPI26156 PI26156
Pierce™ Agarose ChIP Kit, Thermo Scientific
Protein Purification Protein Purification Kits
The Thermo Scientific Pierce Agarose ChIP Kit provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.
  • Simple and fast ChIP assay protocol (< 8 hours)
  • Highly efficient isolation and lysis of nuclei
  • Easy and reproducible enzymatic digestion
  • Low-background and high-binding capacity Protein A/G agarose resin
  • Highly specific positive controls included: RNA polymerase II antibody and GAPDH PCR primers
  • Fast and reproducible spin-column format
  • High-recovery DNA purification
  • ChIP-validated antibodies available
ChIP assay has the ability to capture a snapshot of specific protein-DNA interactions as they occur in living cells, and then quantitate the interactions using standard or quantitive PCR.
Delivery information: Kit includes ChIP Grade Protein A/G Plus Agarose (0.65 mL), IP Dilution/Wash Buffer, 5X, (11 mL), IP Wash Buffer 3, 5X, (4.5 mL), IP Elution Buffer, 2X, (4.5 mL), Column Accessory Pack (3 packs), Microcentrifuge Tubes (1.5mL), 75 tubes, DNA Clean-Up Column (40 columns), DNA Column Binding Solution (30 mL), DNA Column Wash Solution (6 mL), pH Indicator (0.8 mL), DNA Column Elution Solution (5 mL), Anti-RNA Polymerase II Antibody (25 µL), Normal Rabbit IgG, 1 mg/mL, (10 µL), ChIP Positive Control Primers, GAPDH promoter, Human-specific, (100 µL), Pierce Chromatin Prep Module.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty