Would you like to log in again?
Pierce™ Immunoprecipitation, HA-Tag IP/Co-IP Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce Pierce™
26180 26180
PI26180EA 806.4 USD InStock
PI26180 CAPI26180
Pierce™ Immunoprecipitation, HA-Tag IP/Co-IP Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ HA-Tag IP/Co-IP kit provides the affinity resin, positive control and other reagents necessary to perform immunoprecipitation (IP) or co-immunoprecipitation (Co-IP) reactions with an HA-tagged bait protein.
  • Highly specific anti-HA monoclonal antibody provides high-yield immunoprecipitation products
  • The kit and anti-HA agarose are compatible with various cell lysates and physiologic buffer systems
This kit is based on crosslinked beaded agarose to which a highly specific anti-HA antibody is covalently immobilised. The ready-to-use affinity resin, together with the included buffers, microcentrifuge spin columns, positive control, and easy-to-follow instructions, constitute a complete set of reagents sufficient for 25 for IP or Co-IP assays.
Delivery information: Kit includes Anti-HA agarose (0.25 ml), HA-tagged positive control (0.5 ml), BupH Tris buffered saline pack (500 ml), Lane Marker Non-reducing Sample Buffer 5X, (5 ml), spin columns accessory pack (27 columns) and collection tubes and caps accessory pack (100 tubes).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty