Would you like to log in again?
RBS™-pF Laboratory Detergent Concentrate, Thermo Scientific
Supplier: Thermo Scientific
RBS™
27959 27959 27960 27960
PI27959EA 91.63 USD InStock
PI27959 CAPI27960 CAPI27959 PI27960
RBS™-pF Laboratory Detergent Concentrate, Thermo Scientific
Cleaning Agents Detergents and Surface Cleansers
General laboratory cleaner is phosphate-free
Permits long soaking and rinses clean. Safe and easy-to-use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty