Would you like to log in again?
Pierce™ Surfact-Amps™ Detergent Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce Pierce™
28336 85114 28316 28313 28328 28313 28230 28324 85113 28316 85115 85113 28328 28332 28321 85111 28320 85114 28332 28314 85115 85112 28321 28324 85111 85118 85125 85124 85117 85124 28329 28230 85118 28329 85117 28320 28314 28336 85112
PI28329EA 195.46 USD InStock
CAPI28320 CAPI28321 CAPI85118 CAPI85117 PI28329 PI28328 CAPI85115 CAPI85114 CAPI28328 PI28324 CAPI28329 CAPI28324 PI28321 PI85125 PI28320 PI85124 PI85118 PI85117 CAPI85124 CAPI28332 PI28316 CAPI28230 PI28336 PI28314 PI28313 PI85111 PI85114 CAPI28313 PI28332 PI85115 CAPI28314 CAPI28336 PI85112 CAPI28316 PI28230 PI85113 CAPI85113 CAPI85112 CAPI85111
Pierce™ Surfact-Amps™ Detergent Solutions, Thermo Scientific
Protein Extraction Protein Extraction Reagents
Surfact-Amps™ detergents are ready-to-use, diluted solutions of purified detergent that can be used in routine and high-demand protein research methods and molecular biology techniques.
  • Clear and pure 10% detergent solutions with <1.0 μeq/ml peroxides and carbonyl content
  • Ampoules are packed under nitrogen for maximum shelf life
  • Pre-scored for easy opening
  • Easy to pipette and accurately dispense
  • Packaging eliminates the accumulation of peroxides and degradation products
Detergents are a class of molecules whose unique properties enable manipulation (disruption or formation) of hydrophobichydrophilic interactions among molecules in biological samples. In biological research, detergents are used to lyse cells (release soluble proteins), solubilize membrane proteins and lipids, control protein crystallization and prevent non specific binding in affinity purification and immunoassay procedures. They are often used as an additive in protein gel electrophoresis. These detergents are useful as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins, improve chromatography, prepare yeast spheroplasts, and reduce background in ELISA or Western blot methods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty