Would you like to log in again?
Pierce™ Wash Buffers, Thermo Scientific
Supplier: Thermo Scientific
Pierce
28344 28348 28352 28348 28341 28344 28358 28341 28346 28346 28358 28360 28352 28360
PI28348EA 179.63 USD InStock
CAPI28344 CAPI28360 PI28348 CAPI28341 CAPI28352 PI28358 PI28346 PI28344 CAPI28346 CAPI28358 CAPI28348 PI28352 PI28341 PI28360
Pierce™ Wash Buffers, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers
These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.
  • Ready to use without having to reconstitute with ultrapure water
  • Eliminates the possibility of powder remaining in a packet
  • 10X or 20X concentration eliminates time spent waiting for powder to dissolve
  • Storage as concentrated stock minimizes bench space needed for large volume solutions
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25˚C.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty