Would you like to log in again?
Pierce BupH™ Dry Blend Buffers, Washing, Thermo Scientific
Supplier: Thermo Scientific
BupH™
28382 28384 28374 28376 28382 28390 28374 28384 28376 28379 28390 28372 28372 28379
PI28379EA 111.42 USD InStock
CAPI28382 CAPI28372 CAPI28384 CAPI28374 PI28379 CAPI28379 PI28374 PI28384 PI28372 PI-28376 PI28382 CAPI-28376 PI28390 CAPI28390
Pierce BupH™ Dry Blend Buffers, Washing, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers
BupH™ packs are pre-blended and pre-measured dry mixtures of buffers that are easy to prepare and used in protein purification and common laboratory techniques.
  • Packs are pouches of dry-blend powder that are each sufficient to make 500 mL of buffer
  • Eliminates weighing time and pH adjustments
  • Every pack will yield the same, consistent buffer
  • Eliminates concerns about long-term stability of stock solutions
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty