Would you like to log in again?
Pierce™ Protein Conjugate, Bovine Serum Albumin, Biotinylated, Thermo Scientific
Supplier: Thermo Scientific
Pierce
29130 29130
PI29130EA 280.43 USD InStock
PI29130 CAPI29130
Pierce™ Protein Conjugate, Bovine Serum Albumin, Biotinylated, Thermo Scientific
Assays Protein Assays
Biotinylated bovine serum albumin (B-BSA) is used as a positive control in biotin quantitation methods.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty