Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ NeutrAvidin™ Agarose, Thermo Scientific

Supplier: Thermo Fisher Scientific
NeutrAvidin™
29202 29204 29201 29204 29200 29200 29201 29202
PI29204EA 858.09 USD InStock
CAPI29200 CAPI29201 CAPI29202 PI29204 PI29202 CAPI29204 PI29201 PI29200
Pierce™ NeutrAvidin™ Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
These NeutrAvidin™ Agarose is a beaded agarose resin of immobilized NeutrAvidin™ Protein, a modified form of avidin with exceptional biotin-binding characteristics for affinity purification methods.

  • Tetrameric with four biotin-binding site per molecule
  • Support is crosslinked 6% beaded agarose (CL-6B)
  • Immobilised by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding

NeutrAvidin™ agarose consists of a specially purified and deglycosylated form of avidin used for variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.

Standard capacity beads: Have a normal load of immobilised NeutrAvidin™ protein, providing a binding capacity of 1 to 2 mg of biotinylated BSA per ml of resin.

High capacity beads: Have a dense load of immobilised NeutrAvidin™ protein, providing a binding capacity greater than 8 mg of biotinylated BSA per ml of resin.

Properties of crosslinked 6% beaded agarose (CL-6B):
Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
Average Particle Size: 45 to 165 microns
Exclusion Limit: 10,000 to 4,000,000 daltons
Maximum Volumetric Flow Rate: approx. 1 mL/minute (for 1 cm diameter column)
Maximum Linear Velocity: 30 cm per hour
Maximum Pressure: less than 25psi (1.5 bar)

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty