Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Recombinant Protein G (from E. coli), Biotin

Supplier: Pierce Chemical
Pierce
29988 29988
PI29988EA 337.29 USD InStock
PI29988 CAPI29988
Pierce™ Recombinant Protein G (from E. coli), Biotin
Proteins and Peptides
Biotinylated recombinant Protein G can be used to probe and detect mouse and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains two Fc-binding domains per protein
  • Does not bind human IgM, IgD or IgA
  • More efficient than protein A for mouse (IgG1), human (IgG3) and rat, goat and cow antibodies
  • Less efficient than protein A for guinea pig, pig, dog and cat

Protein G is a bacterial cell wall protein isolated from group G Streptococci. DNA sequencing of native protein G identifies two IgG-binding domains and sites for albumin and cell surface binding. The albumin and cell surface binding domains have been eliminated from recombinant protein G to reduce non specific binding and, therefore, can be used to separate IgG from crude samples. Optimal binding occurs at pH 5, although binding is also effective at pH 7,0 to 7,2.

Form: salt-free powder
A280 of 0.1% solution: 1.0
Isoelectric point (pI): 4.5

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty