Would you like to log in again?
Pierce™ Avidin, HRP (Horseradish Peroxidase)
Supplier: Pierce Chemical
Pierce
29994 21123 29994
PI29994EA 485.91 USD InStock
CAPI29994 PI29994 CAPI21123
Pierce™ Avidin, HRP (Horseradish Peroxidase)
Proteins and Peptides
Pierce™ HRP conjugated avidin is used in biotin-based detection methods, including Western blotting or ELISA detection with appropriate enzyme substrates.
  • Native avidin - glycoprotein from chicken egg whites, 67 kDa, pI = 10
  • Glycosylation and isoelectric point renders avidin more soluble than streptavidin
  • Significantly less expensive than recombinant streptavidin
  • Avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
  • HRP conjugate ideal for membrane and plate-based assays
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty