Would you like to log in again?
Pierce™ High Sensitivity NeutrAvidin™ HRP
Supplier: Pierce Chemical
Pierce
31030 31030 31032
PI31032EA 1421.54 USD InStock
PI31032 CAPI31030 PI31030
Pierce™ High Sensitivity NeutrAvidin™ HRP
Proteins and Peptides
These High Sensitivity NeutrAvidin HRP Conjugate is specially prepared peroxidase-conjugated form of avidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.
  • High specificity, low background binding
  • High sensitivity which detects low levels of target without background; obtain high signal-to-noise ratios
  • Cost effective, uses less conjugate in Western blotting and ELISA applications while still obtaining excellent results
  • Compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
  • Ready-to-use stabilised liquid format means there is no waiting for thawing and no need to aliquot
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty