Would you like to log in again?
Pierce™ EZ-Link™ Maleimide Activated Horseradish Peroxidase Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
31485 31494 31485 31494
PI31485EA 351.83 USD InStock
PI31485 PI31494 CAPI31485 CAPI31494
Pierce™ EZ-Link™ Maleimide Activated Horseradish Peroxidase Kit, Thermo Scientific
Protein Linking and Labeling Systems
Thermo Scientific Pierce Maleimide Activated Horseradish Peroxidase is for preparation of HRP conjugates with proteins, peptides or other ligands that contain sulfhydryl groups, such as reduced cysteines.
  • Activated HRP
  • Sulfhydryl-reactive
  • Product is shipped at ambient temperature
This kit contains 5 mg of conjugated protein and can effectively modify 5 mg of immunoglobulin and also contains all buffers and one 10 mL desalting column.
Total weight is
25mg as a result of buffer components and salts lyophilized along with the activated protein.
Delivery information: Kit contains two modification reagents allowing flexibility in the method of producing free sulfhydryls on macromolecules. 2-MercaptoethylamineHCl is a mild reagent for reduction of IgG and F(ab´)2 fragments. SATA is a sulfhydryl-containing modification reagent that reacts with primary amines to present protected sulfhydryl groups, which can be exposed upon treatment with hydroxylamine.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty