Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pierce™ ECL Western Blotting Substrate, Thermo Scientific
Supplier: Thermo Scientific
Pierce
32109 32209 32109 32106
PI32106EA 302.4 USD InStock
PI32106 PI32109 PI32209 CAPI32109
Pierce™ ECL Western Blotting Substrate, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
A value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.
  • Less cost than other ECL Substrates
  • Switch from other entry-level ECL substrates without the need for optimization or protocol changes
This substrate provides reliability and performance equivalent to other standard ECL substrates for detection of horseradish peroxidase (HRP) enzyme activity. The luminol and peroxide reagent-formulations are identical to other available substrate products, so one can switch to this economical substrate without needing to optimize probing conditions or incubation protocols.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty