Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Recombinant Protein A/G (from E. coli)

Supplier: Pierce Chemical
Pierce
32391 32391
PI32391EA 387.69 USD InStock
PI32391 CAPI32391
Pierce™ Recombinant Protein A/G (from E. coli)
Proteins and Peptides
Pierce™ Recombinant Alkaline Phosphatase Conjugated Protein A/G is useful to probe and detect antibodies, especially IgG isotypes, of many different species hosts in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains four Fc-binding domains from Protein A and two from Protein G
  • Provides binding for all antibody species and subclasses recognised by either Protein A or Protein G

The extended Fc-binding properties of Protein A/G make it a tool in the investigation and purification of immunoglobulins. Protein A/G binds to all human IgG subclasses, IgA, IgE, IgM and to a lesser extent IgD; however, it does not bind mouse IgA, IgM or murine serum albumin. Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins. Individual subclasses of mouse monoclonal are most likely to have stronger affinity to this chimeric protein than to either Protein A or Protein G.

Form: salt-free powder
A280 of 0.1% solution:

0.505
Isoelectric point (pI): 4.65

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty