Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Recombinant Protein A (from E. coli)

Supplier: Pierce Chemical
Pierce
32400 32400
PI32400EA 261.05 USD InStock
CAPI32400 PI32400
Recombinant Protein A (from E. coli)
Proteins and Peptides
Pierce™ Recombinant Peroxidase Conjugated (HRP-Horseradish Peroxidase) Protein A can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains four Fc-binding domains per protein
  • More efficient than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
  • Less efficient than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies

Protein A is a cell wall component produced by several strains of Staphylococcus aureus that consists of a single polypeptide chain and contains little or no carbohydrate. Recombinant Protein A is produced in E. coli and functions essentially the same as native Protein A. Contains four high-affinity binding sites capable of interacting with the Fc region from IgG of several species including human and rabbit. Optimal binding occurs at pH 8.2, although binding is also effective at neutral or physiological conditions (pH 7.0 to 7.6)

Form: salt-free powder
A280 of 0.1% solution:

0.165
Isoelectric point (pI): 4.7 to 4.8

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty