Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Recombinant Protein L (from E. coli)

Supplier: Pierce Chemical
Pierce
32420 32420
PI32420EA 319.2 USD InStock
PI32420 CAPI32420
Pierce™ Recombinant Protein L (from E. coli)
Proteins and Peptides
Pierce™ Recombinant Peroxidase Conjugated (HRP-Horseradish Peroxidase) Protein L can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains four immunoglobulin-binding domains per protein
  • Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
  • Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Form: salt-free powder
A280 of 0.1% solution:

0.80
Isoelectric point (pI): 4.8

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty