Would you like to log in again?
Pierce™ HRP Substrates, 1-Step™ Turbo TMB-ELISA, Thermo Scientific
Supplier: Thermo Scientific
Pierce
34022 34022
PI34022EA 214.8 USD InStock
CAPI34022 PI34022
Pierce™ HRP Substrates, 1-Step™ Turbo TMB-ELISA, Thermo Scientific
Assays ELISAs
TMB (3,3´,5,5´ tetramethylbenzidine) is a substrate for horseradish peroxidase (HRP).
The 1-Step™ Turbo TMB-ELISA is a complete solution that contains TMB and a stabilised hydrogen peroxide. TMB has an absorption maximum at 285 nm before oxidation. After oxidation, the maxima are 370 and 652 nm. The blue solution can be measured at 652 nm. However, upon addition of the stop solution (ie. acid) a yellow solution results that can be measured at 450 nm and has a higher sensitivity than the measurement of the blue solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty