Would you like to log in again?
Pierce™ 1-Step™ Ultra TMB-ELISA Substrate Solution, Thermo Scientific
Supplier: Thermo Scientific
Pierce®
34029 34029 34028 34028
PI34029EA 758.4 USD InStock
PI34029 PI34028 CAPI34028 CAPI34029
Pierce™ 1-Step™ Ultra TMB-ELISA Substrate Solution, Thermo Scientific
Assays ELISAs
The 1-Step™ Ultra TMB-ELISA contains soluble TMB (3,3´,5,5´-tetramentylbenzidine) that is ideal for sensitive ELISA-based applications
  • Available in traditional or ready-to-use formulations
  • No additional reagents or filtering required
  • Non carcinogenic
  • Upon receipt store at 4˚C, protected from moisture
  • Product is shipped at ambient temperature
TMB is a chromogenic substrate for horseradish peroxidase. Once oxidised by the enzyme, this substrate yields a blue product that absorbs at 370 nm and 652 nm. Adding sulfuric or phosphoric acid to stop the reaction changes the product to yellow that absorbs at 450 nm and provides greater sensitivity than the blue product. The 1-Step Ultra TMB-ELISA is supplied ready to use with no mixing required.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty