Would you like to log in again?
Pierce™ ONPG (o-nitrophenyl-β-D-galactopyranoside), Thermo Scientific
Supplier: Thermo Scientific
Pierce®
34055 34055
PI34055EA 154.51 USD InStock
CAPI34055 PI34055
Pierce™ ONPG (o-nitrophenyl-β-D-galactopyranoside), Thermo Scientific
Assays ELISAs
Thermo Scientific Pierce ONPG is the preferred colorimetric substrate for ELISA applications involving beta-galactosidase (b-Gal) as the reporter enzyme.
  • Product formed is completely soluble
  • High extinction coefficient at 405nm
  • Substrate yields a yellow product that is easily detectable in the visual range after stopping the reaction with 1M sodium carbonate
When using beta-Galactosidase as the label for proteins in ELISA studies, a wide variety of substrates are available, including: o-Nitrophenyl-beta-D-galactopyranosidase (ONPG), Naphthol-AS-Bl-beta-D-galactopyranosidase (Nap-Gal), and 4-Methyl-umbelliferyl-beta-D-galactopyranosidase (MUm-Gal). However, it is important to choose a substrate with adequate solubility, that uses readily available equipment, and that gives a significant reading over the background. ONPG is a superior beta-Galactosidase substrate option. The product formed is completely soluble and has a high extinction coefficient at 405nm. The substrate yields a yellow product that is easily detectable in the visual range after stopping the reaction with 1M Sodium Carbonate.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty