Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SuperSignal™ West Pico Complete Mouse IgG Detection Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce
34084 34083 34082 34083 34081 34084 34081 34082
PI34081EA 652.62 USD InStock
CAPI34081 CAPI34082 CAPI34083 CAPI34084 PI34081 PI34084 PI34083 PI34082
SuperSignal™ West Pico Complete Mouse IgG Detection Kit, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
Thermo Scientific SuperSignal West Pico Complete IgG Detection Kit is a a kit containing goat anti-mouse or anti-rabbit secondary antibody HRP conjugate for the detection of mouse or rabbit primary antibody
  • Optimized - secondary antibodies are validated for use with the SuperSignal West Pico Chemiluminescent Substrate, eliminating the need to test multiple dilutions
  • Large packages - kits provide sufficient reagents to complete up to 50 experiments with standard mini gel blots (4000 square centimeters of membrane)
  • Standard or complete - choose a kit with substrate and antibody only, or a complete kit that also includes TBS wash buffer and high-performance SuperBlock Blocking Buffer
All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate. The kits are offered in a standard format (substrate and optimized HRP-conjugates) and a complete format that also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.
The kit is ideal for busy researchers or new beginners to Western blot analysis.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty