Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Pierce™ SuperSignal™ West Pico PLUS Chemiluminescent Substrate, Thermo Scientific

Supplier: Thermo Scientific
Pierce
34578 34579 34577 34580
PI34577EA 205.69 USD InStock
PI34577 PI34579 PI34578 PI34580
Pierce™ SuperSignal™ West Pico PLUS Chemiluminescent Substrate, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
Thermo Scientific SuperSignal™ West Pico PLUS Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) HRP substrate for picogram to high-femtogram level detection.

  • Picogram to Femtogram sensitivity: Detect low-picogram to high-femtogram amounts of target protein on nitrocellulose or PVDF membrane
  • Long signal duration: Incubated blots provide up to 24 hours of usable light output when conditions are optimized
  • Stable reagent: Eight hour working solution stability; One-year kit stability at room temperature

SuperSignal West Pico PLUS Substrate provides excellent signal intensity and sensitivity for western blotting with horseradish peroxidase (HRP) enzyme conjugates. The intensity of the light emission combined with the exceptional duration allows for acquiring multiple exposures to effortlessly obtain publication-quality blot images. The chemiluminescent substrate is compatible with different membranes, blocking reagents and a wide range of antibody dilutions making it an ideal choice for most western blotting applications.

Provides high performance outside of the recommended antibody dilution ranges, including the most common 1:5 K to 1:10 K secondary antibody dilutions of a 1 mg/ml stock solution.

Optimized for dilute antibody concentrations:
0.2-1.0 ug/mL primary antibody (1:1000 to 1:5000 dilution from 1 mg/mL stock)
10-50 ng/mL secondary antibody (1:20,000 to 1:100,000 dilution from 1 mg/mL stock)

Optimized for dilute antibody concentrations:
0.2 to 1.0 ug/mL primary antibody (1:1000 to 1:5000 dilution from 1 mg/mL stock)
10 to 50 ng/mL secondary antibody (1:20,000 to 1:100,000 dilution from 1 mg/mL stock)

The intensity of the light emission combined with the exceptional duration allows for acquiring multiple exposures to effortlessly obtain publication-quality blot images. The chemiluminescent substrate is compatible with different membranes, blocking reagents and a wide range of antibody dilutions making it an ideal choice for most western blotting applications.

Delivery information: Kit contains SuperSignal™ West Pico PLUS Luminol/Enhancer (10, 100, 250 or 500 mL, depending on pack size) and SuperSignal™ West Pico PLUS Stable Peroxide (10, 100, 250 or 500 mL, depending on pack size).

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty