Would you like to log in again?
Pierce™ Fast Western Blot Kits, SuperSignal™ West Dura, Mouse, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
35070 35071 35070 35071
PI35071EA 558.28 USD InStock
CAPI35071 CAPI35070 PI35071 PI35070
Pierce™ Fast Western Blot Kits, SuperSignal™ West Dura, Mouse, Thermo Scientific
Blotting Kits
The Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate provides reagents and protocol for reliable and rapid, 1-hour chemiluminescent Western blotting at femtogram levels of sensitivity.
  • Fast—femtogram levels of detection in about 1 hour
  • Convenient—no expensive hardware or vacuum required; no clogging issues
  • Simple—optimized protocol makes Western blot analysis easier than ever
  • Economical—no expensive consumables or extra equipment
  • Excellent stability—kit is stable for 1 year stored at 4 °C
  • Easy—complete kits contain all components needed to block, probe and develop a blot with your mouse primary antibody
The Fast Western Blot Kit protocol is a quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other rapid Western blotting systems. These complete Pierce Fast Western Blot Kits require minimal hands-on time and yield results comparable to classical Western blotting with SuperSignal West Dura Chemiluminescent Substrate. Like other Western blotting methods, the included SuperSignal West Dura Substrate produces a signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed. The complete kit contains all the reagents necessary to block and probe a Western blot with mouse primary antibody.
Delivery information: Kit includes Fast Western antibody diluent, Fast Western 10X wash buffer, Fast Western (mouse or rabbit) optimised HRP reagent, SuperSignal™ West Dura luminol/enhancer solution, SuperSignal™ West stable peroxide solution.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty