Would you like to log in again?
Pierce SuperSignal™ ELISA Pico Chemiluminescent Substrate, Thermo Scientific
Supplier: Thermo Scientific
Pierce
37069 37069 37070 37070
PI37070EA 429.6 USD InStock
PI37070 CAPI37069 PI37069 CAPI37070
Pierce SuperSignal™ ELISA Pico Chemiluminescent Substrate, Thermo Scientific
Assays ELISAs
SuperSignal™ ELISA Pico chemiluminescent substrate is optimized to generate an intense light signal and provide exceptional performance in luminometer based assays.
  • Intense signal is produced immediately at room temperature or at 37 °C
  • Signal can be read in black or white opaque microplates
  • Detects protein in ELISAs down to the picogram levels
  • Ambient shipping and room temperature storage
  • Consistent performance of the working solution over eight hour period
It offers greater sensitivity in ELISAs, or any other solution-based assay using chemiluminescent substrates for enzyme detection and quantification. These ELISAs can take place in either a test tube or a microplate and are quantified by measuring relative light units (RLU) in a luminometer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty