Would you like to log in again?
Pierce SuperSignal™ ELISA Femto Substrate, Thermo Scientific
Supplier: Thermo Scientific
Pierce
37074 37075 37074 37075
PI37074EA 797.29 USD InStock
CAPI37074 CAPI37075 PI37074 PI37075
Pierce SuperSignal™ ELISA Femto Substrate, Thermo Scientific
Assays ELISAs
SuperSignal® ELISA Femto maximum sensitivity substrate utilizes the SuperSignal® system with an enhancer formulation for superior protein detection and low-end linearity in ELISA applications.
  • Intense signal generated immediately at both room temperature and 37 °C
  • Easy detection of low quantities of proteins with high signal:noise ratios and low-end linearity of dose response curves
  • Femtogram-level detection of target proteins in ELISA
  • High sensitivity allows for reduction in ELISA incubation steps and hence a reduction in assay time
  • Storage at room temperature with a 6 hour working solution stability
  • Ideal for automated high-throughput screening applications
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty