Would you like to log in again?
Pierce™ Blocker™ BSA Blocking Buffer, Thermo Scientific
Supplier: Thermo Scientific
Pierce
37520 37525 37525 37520
PI37525EA 253.51 USD InStock
PI37525 CAPI37520 PI37520 CAPI37525
Pierce™ Blocker™ BSA Blocking Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Blocker BSA in PBS or TBS are ready-to-use, 1X PBS or TBS solutions of 10X (10%) bovine serum albumin protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.
  • Easy to use
  • Convenient and saves storage space
  • Purified protein provides fewer chances of cross reaction with assay components
This 10% (w/v) solution of high quality BSA that is useful for saturating excess protein binding sites on membranes and microplates in immunoassays. Blocker BSA Buffers are most frequently diluted 10-fold (to 1% BSA) in 1X Tris-buffered saline for initial testing, but other buffer concentrations can be beneficial for specific systems. Blocker BSA is usually more effective than non fat milk blocking buffers for biotin-avidin systems because it contains a single purified protein that is devoid endogenous biotin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty