Would you like to log in again?
Pierce™ Fish Serum Blocking Buffe, Thermo Scientific
Supplier: Thermo Scientific
37527 37527
PI37527EA 293.78 USD InStock
PI37527 CAPI37527
Pierce™ Fish Serum Blocking Buffe, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
This Fish Serum Blocking Buffer is Steelhead Salmon Serum in PBS that is especially useful as a blocking agent in IHC and other detection methods involving mammalian samples.
  • Non mammalian
  • Easy to use
  • Convenient, filtered and stabilized in PBS
Fish Serum Blocking Buffer is fish serum, it is effective in stabilizing solutions and samples for antibody-binding interactions while at the same time minimizing the possibility of cross-reaction with mammalian sample components
The blocker has been used to decrease background and increase signal-to-noise ratios with near IR fluorescent probes and is validated for use with the LI-COR Odyssey™ Infrared Imaging System.
The product is also especially effective in various cellular imaging protocols based on visible and near infrared fluorescence detection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty