Would you like to log in again?
SuperBlock™ T20 (TBS) Blocking Buffer, Thermo Scientific
Supplier: Thermo Scientific
Pierce
37535 37537 37537 37536 37581 37581 37535 37536
PI37537EA 266.54 USD InStock
PI37537 CAPI37581 PI37535 PI37536 CAPI37537 PI37581 CAPI37535 CAPI37536
SuperBlock™ T20 (TBS) Blocking Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Thermo Scientific SuperBlock™ Blocking Buffer is optimised solutions in PBS or TBS containing a protein that provides incredibly fast and effective blocking for ELISA, IHC and Western blot analysis.
  • Fast - block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Flexible - guaranteed to be biotin-free for use with streptavidin system
  • Low background - the non-serum protein solution yields a high signal-to-noise ratio
  • Stable - store buffer at 4˚C for one year; store blocked plates dry for up to 12 months
  • Product is shipped at ambient temperature
Blocking microplates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C. Product is shipped at ambient temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty