Would you like to log in again?
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Supplier: Thermo Scientific
Pierce
37578 37578 37539 37542 37543 37538 37579 37543 37542 37538 37539 37579
PI37538EA 299.71 USD InStock
PI37538 PI37539 CAPI37539 PI37578 PI37579 CAPI37578 CAPI37542 PI37542 CAPI37543 PI37543 CAPI37538 CAPI37579
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
StartingBlock Blocking Buffers performs well with an extremely wide range of antibodies, antibody combinations, other protein probing and assay systems.
These buffers are biotin- and serum-free. They are compatible with many detection systems and feature short blocking times. These buffers also allow stripping and reprobing with reblocking to save time and money.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty